Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGAAAATGTTTTTTTCAGTTGTA[C/T]ATTGTTCCAGGGCAAATACTGTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606140 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
XPO7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
XPO7 - exportin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015024.4 | Intron | NP_055839.3 | ||||
XM_017013244.1 | Intron | XP_016868733.1 | ||||
XM_017013245.1 | Intron | XP_016868734.1 | ||||
XM_017013246.1 | Intron | XP_016868735.1 | ||||
XM_017013247.1 | Intron | XP_016868736.1 |
Set Membership: |
HapMap |