Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGGCCAGGAAGTTCTTTTTCAT[A/G]TTCTGGGCTCAATCGTGATGCTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602232 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNQ3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNQ3 - potassium voltage-gated channel subfamily Q member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204824.1 | 11097 | UTR 3 | NP_001191753.1 | |||
NM_004519.3 | 11097 | UTR 3 | NP_004510.1 | |||
XM_005250914.3 | 11097 | UTR 3 | XP_005250971.1 | |||
XM_006716555.3 | 11097 | UTR 3 | XP_006716618.1 | |||
XM_011517026.2 | 11097 | UTR 3 | XP_011515328.1 | |||
XM_017013400.1 | 11097 | UTR 3 | XP_016868889.1 |
Set Membership: |
HapMap |