Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614930 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LRRC6 - leucine rich repeat containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321961.1 | 2154 | Missense Mutation | ACT,ATT | T,I 446 | NP_001308890.1 | |
NM_001321962.1 | 2154 | Missense Mutation | ACT,ATT | T,I 384 | NP_001308891.1 | |
NM_001321963.1 | 2154 | Missense Mutation | ACT,ATT | T,I 346 | NP_001308892.1 | |
NM_001321964.1 | 2154 | Missense Mutation | ACT,ATT | T,I 346 | NP_001308893.1 | |
NM_001321965.1 | 2154 | Missense Mutation | ACT,ATT | T,I 346 | NP_001308894.1 | |
NM_001321966.1 | 2154 | Missense Mutation | ACT,ATT | T,I 326 | NP_001308895.1 | |
NM_012472.5 | 2154 | Missense Mutation | ACT,ATT | T,I 466 | NP_036604.2 | |
XM_006716538.3 | 2154 | Missense Mutation | ACT,ATT | T,I 472 | XP_006716601.2 | |
XM_011516950.2 | 2154 | Missense Mutation | ACT,ATT | T,I 452 | XP_011515252.1 | |
XM_017013296.1 | 2154 | Missense Mutation | ACT,ATT | T,I 432 | XP_016868785.1 | |
XM_017013297.1 | 2154 | Missense Mutation | ACT,ATT | T,I 346 | XP_016868786.1 | |
XM_017013298.1 | 2154 | Missense Mutation | ACT,ATT | T,I 346 | XP_016868787.1 |