Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCATTCATTTAATCTTTACAACC[A/C]TACAAACACGCATTACAATTATGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604331 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTS6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
INTS6 - integrator complex subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039937.1 | 4359 | Intron | NP_001035026.1 | |||
NM_001039938.1 | 4359 | Intron | NP_001035027.1 | |||
NM_001306091.1 | 4359 | Intron | NP_001293020.1 | |||
NM_012141.2 | 4359 | Intron | NP_036273.1 | |||
XM_011535040.2 | 4359 | UTR 3 | XP_011533342.1 | |||
XM_011535041.1 | 4359 | Intron | XP_011533343.1 | |||
XM_017020540.1 | 4359 | Intron | XP_016876029.1 |
MIR5693 - microRNA 5693 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERPINE3 - serpin family E member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101320.1 | 4359 | Intron | NP_001094790.1 | |||
XM_017020707.1 | 4359 | Intron | XP_016876196.1 |
Set Membership: |
HapMap |