Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACATTCTGAGATTTTTTTGCAAC[G/T]TTTTTAGCTTACCAGCGATTGTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165070 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FLT1 - fms related tyrosine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159920.1 | Intron | NP_001153392.1 | ||||
NM_001160030.1 | Intron | NP_001153502.1 | ||||
NM_001160031.1 | Intron | NP_001153503.1 | ||||
NM_002019.4 | Intron | NP_002010.2 | ||||
XM_011535014.1 | Intron | XP_011533316.1 | ||||
XM_017020485.1 | Intron | XP_016875974.1 |
Set Membership: |
HapMap |