Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610348 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLF2 - SMC5-SMC6 complex localization factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136123.1 | 738 | Intron | NP_001129595.1 | |||
NM_001243770.1 | 738 | UTR 3 | NP_001230699.1 | |||
NM_018121.3 | 738 | Intron | NP_060591.3 | |||
XM_005269965.2 | 738 | Intron | XP_005270022.1 | |||
XM_011539944.2 | 738 | UTR 5 | XP_011538246.1 |