Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTAGAAAAGTCCTCTTGCTCCA[C/T]TCAGTTGTTCTCCACACTTTAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600763 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNORA31 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNORA31 - small nucleolar RNA, H/ACA box 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPT1 - tumor protein, translationally-controlled 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286272.1 | 3267 | UTR 3 | NP_001273201.1 | |||
NM_001286273.1 | 3267 | UTR 3 | NP_001273202.1 | |||
NM_003295.3 | 3267 | UTR 3 | NP_003286.1 |
TPT1-AS1 - TPT1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |