Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACTTTGCATTTGCAGAGAATGTT[A/G]CAGAATTTCAAAGTATTTCAAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
35 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608756 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COLGALT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COLGALT2 - collagen beta(1-O)galactosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSEN15 - tRNA splicing endonuclease subunit 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127394.3 | Intron | NP_001120866.1 | ||||
NM_001300764.1 | Intron | NP_001287693.1 | ||||
NM_001300766.1 | Intron | NP_001287695.1 | ||||
NM_052965.3 | Intron | NP_443197.1 | ||||
XM_006711148.1 | Intron | XP_006711211.1 | ||||
XM_011509139.2 | Intron | XP_011507441.1 | ||||
XM_017000228.1 | Intron | XP_016855717.1 | ||||
XM_017000229.1 | Intron | XP_016855718.1 | ||||
XM_017000230.1 | Intron | XP_016855719.1 |