Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
49 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608375 MIM: 601007 MIM: 613461 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DNAJC6 - DnaJ heat shock protein family (Hsp40) member C6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LEPR - leptin receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003679.3 | Intron | NP_001003679.1 | ||||
NM_001003680.3 | Intron | NP_001003680.1 | ||||
NM_001198687.1 | Intron | NP_001185616.1 | ||||
NM_001198688.1 | Intron | NP_001185617.1 | ||||
NM_001198689.1 | Intron | NP_001185618.1 | ||||
NM_002303.5 | Intron | NP_002294.2 |
LEPROT - leptin receptor overlapping transcript | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198681.1 | Intron | NP_001185610.1 | ||||
NM_001198683.1 | Intron | NP_001185612.1 | ||||
NM_017526.4 | Intron | NP_059996.1 |
Set Membership: |
HapMap JSNP Validated |