Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGTAATGGAGACAATACTTCACC[A/G]TGTTTCCAAAATATTTTACAGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605439 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EHF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EHF - ETS homologous factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206615.1 | Intron | NP_001193544.1 | ||||
NM_001206616.1 | Intron | NP_001193545.1 | ||||
NM_012153.5 | Intron | NP_036285.2 | ||||
XM_005252857.1 | Intron | XP_005252914.1 | ||||
XM_005252860.1 | Intron | XP_005252917.1 | ||||
XM_005252861.1 | Intron | XP_005252918.1 | ||||
XM_005252862.1 | Intron | XP_005252919.1 | ||||
XM_011519983.1 | Intron | XP_011518285.1 | ||||
XM_011519984.1 | Intron | XP_011518286.1 | ||||
XM_011519985.1 | Intron | XP_011518287.1 | ||||
XM_017017534.1 | Intron | XP_016873023.1 |