Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610661 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NGLY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
NGLY1 - N-glycanase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145293.1 | Intron | NP_001138765.1 | ||||
NM_001145294.1 | Intron | NP_001138766.1 | ||||
NM_001145295.1 | Intron | NP_001138767.1 | ||||
NM_018297.3 | Intron | NP_060767.2 | ||||
XM_005265316.1 | Intron | XP_005265373.1 | ||||
XM_005265317.1 | Intron | XP_005265374.1 | ||||
XM_011533944.1 | Intron | XP_011532246.1 | ||||
XM_011533945.2 | Intron | XP_011532247.1 | ||||
XM_017006839.1 | Intron | XP_016862328.1 |
Set Membership: |
HapMap Validated |