Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCTTAGATGAAATTTGAGCAGA[G/T]ACGTGAATGTATGTGGGACATAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614442 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PSD4 - pleckstrin and Sec7 domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012455.2 | Intron | NP_036587.2 | ||||
XM_005263634.2 | Intron | XP_005263691.1 | ||||
XM_006712392.2 | Intron | XP_006712455.1 | ||||
XM_006712393.2 | Intron | XP_006712456.1 | ||||
XM_006712394.2 | Intron | XP_006712457.1 |
Set Membership: |
HapMap |