Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGGTGAATTTTCTTTTCTCCTT[A/G]TGCATATTTTTCCCCTTAAACATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609451 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP90 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZFP90 - ZFP90 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305203.1 | Intron | NP_001292132.1 | ||||
NM_001305204.1 | Intron | NP_001292133.1 | ||||
NM_001305206.1 | Intron | NP_001292135.1 | ||||
NM_001305207.1 | Intron | NP_001292136.1 | ||||
NM_001305208.1 | Intron | NP_001292137.1 | ||||
NM_133458.3 | Intron | NP_597715.2 | ||||
XM_005255804.2 | Intron | XP_005255861.1 | ||||
XM_017022952.1 | Intron | XP_016878441.1 |
Set Membership: |
HapMap |