Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604813 MIM: 605823 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX17 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
COX17 - COX17, cytochrome c oxidase copper chaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005694.1 | 461 | Intron | NP_005685.1 |
POPDC2 - popeye domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308333.1 | 461 | Silent Mutation | CGC,CGT | R,R 98 | NP_001295262.1 | |
NM_022135.3 | 461 | Silent Mutation | CGC,CGT | R,R 98 | NP_071418.2 | |
XM_005247698.3 | 461 | Silent Mutation | CGC,CGT | R,R 98 | XP_005247755.1 |
Set Membership: |
HapMap Validated |