Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTAGAGGTATCTCATTCTAAGGA[A/G]ATGCGAGTGGGAAGCTGGGTCGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601774 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGAT5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MGAT5 - mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002410.4 | Intron | NP_002401.1 | ||||
XM_005263668.4 | Intron | XP_005263725.1 | ||||
XM_005263669.4 | Intron | XP_005263726.1 | ||||
XM_005263670.3 | Intron | XP_005263727.1 | ||||
XM_006712534.3 | Intron | XP_006712597.1 | ||||
XM_011511199.2 | Intron | XP_011509501.1 | ||||
XM_011511200.1 | Intron | XP_011509502.1 | ||||
XM_011511201.2 | Intron | XP_011509503.1 | ||||
XM_011511202.1 | Intron | XP_011509504.1 | ||||
XM_011511204.2 | Intron | XP_011509506.1 | ||||
XM_017004147.1 | Intron | XP_016859636.1 | ||||
XM_017004148.1 | Intron | XP_016859637.1 | ||||
XM_017004149.1 | Intron | XP_016859638.1 |
MIR3679 - microRNA 3679 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |