Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608170 MIM: 611435 MIM: 605903 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX41 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DDX41 - DEAD-box helicase 41 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DOK3 - docking protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144875.1 | 1740 | Intron | NP_001138347.1 | |||
NM_001144876.1 | 1740 | Intron | NP_001138348.1 | |||
NM_001308235.1 | 1740 | Intron | NP_001295164.1 | |||
NM_001308236.1 | 1740 | Intron | NP_001295165.1 | |||
NM_024872.2 | 1740 | Intron | NP_079148.2 | |||
XM_005265985.3 | 1740 | UTR 3 | XP_005266042.1 | |||
XM_005265987.1 | 1740 | Intron | XP_005266044.1 | |||
XM_011534654.2 | 1740 | UTR 3 | XP_011532956.1 | |||
XM_011534655.2 | 1740 | UTR 3 | XP_011532957.1 | |||
XM_011534656.2 | 1740 | UTR 3 | XP_011532958.1 | |||
XM_011534657.2 | 1740 | UTR 3 | XP_011532959.1 | |||
XM_017009883.1 | 1740 | UTR 3 | XP_016865372.1 | |||
XM_017009884.1 | 1740 | Intron | XP_016865373.1 |
PDLIM7 - PDZ and LIM domain 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |