Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF558 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF558 - zinc finger protein 558 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304350.1 | 3248 | UTR 3 | NP_001291279.1 | |||
NM_144693.2 | 3248 | UTR 3 | NP_653294.1 | |||
XM_011527703.2 | 3248 | UTR 3 | XP_011526005.1 | |||
XM_017026370.1 | 3248 | UTR 3 | XP_016881859.1 | |||
XM_017026371.1 | 3248 | UTR 3 | XP_016881860.1 | |||
XM_017026372.1 | 3248 | UTR 3 | XP_016881861.1 |
Set Membership: |
HapMap |