Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
42 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610378 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLIS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GLIS1 - GLIS family zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_147193.2 | 2237 | UTR 3 | NP_671726.2 | |||
XM_017000408.1 | 2237 | UTR 3 | XP_016855897.1 | |||
XM_017000409.1 | 2237 | UTR 3 | XP_016855898.1 | |||
XM_017000410.1 | 2237 | UTR 3 | XP_016855899.1 | |||
XM_017000411.1 | 2237 | UTR 3 | XP_016855900.1 | |||
XM_017000412.1 | 2237 | Intron | XP_016855901.1 |
Set Membership: |
JSNP Validated |