Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCATACTATAAACATTATTTGGCC[A/G]GGCACAGTGGCACACGCCTAGAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 166945 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929767 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929767 - uncharacterized LOC101929767 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NBR1 - NBR1, autophagy cargo receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291571.1 | Intron | NP_001278500.1 | ||||
NM_001291572.1 | Intron | NP_001278501.1 | ||||
NM_005899.4 | Intron | NP_005890.2 | ||||
NM_031862.3 | Intron | NP_114068.1 | ||||
XM_006721903.1 | Intron | XP_006721966.1 | ||||
XM_011524813.1 | Intron | XP_011523115.1 | ||||
XM_017024642.1 | Intron | XP_016880131.1 | ||||
XM_017024643.1 | Intron | XP_016880132.1 | ||||
XM_017024644.1 | Intron | XP_016880133.1 |