Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604613 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBX18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBX18 - T-box 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080508.2 | Intron | NP_001073977.1 | ||||
XM_006715602.3 | Intron | XP_006715665.2 | ||||
XM_011536247.2 | Intron | XP_011534549.2 | ||||
XM_011536248.2 | Intron | XP_011534550.2 | ||||
XM_011536249.2 | Intron | XP_011534551.2 | ||||
XM_017011472.1 | Intron | XP_016866961.1 | ||||
XM_017011473.1 | Intron | XP_016866962.1 |