Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604613 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBX18 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TBX18 - T-box 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080508.2 | Intron | NP_001073977.1 | ||||
XM_006715602.3 | Intron | XP_006715665.2 | ||||
XM_011536247.2 | Intron | XP_011534549.2 | ||||
XM_011536248.2 | Intron | XP_011534550.2 | ||||
XM_011536249.2 | Intron | XP_011534551.2 | ||||
XM_017011472.1 | Intron | XP_016866961.1 | ||||
XM_017011473.1 | Intron | XP_016866962.1 |
Set Membership: |
HapMap Validated |