Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAAACTGCCAGAGATAAACACAT[A/T]ATGAAGAAAAGGAGAGGCAACTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614605 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZDHHC21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZDHHC21 - zinc finger DHHC-type containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178566.4 | Intron | NP_848661.1 | ||||
XM_017014682.1 | Intron | XP_016870171.1 | ||||
XM_017014683.1 | Intron | XP_016870172.1 | ||||
XM_017014684.1 | Intron | XP_016870173.1 | ||||
XM_017014685.1 | Intron | XP_016870174.1 | ||||
XM_017014686.1 | Intron | XP_016870175.1 | ||||
XM_017014687.1 | Intron | XP_016870176.1 | ||||
XM_017014688.1 | Intron | XP_016870177.1 | ||||
XM_017014689.1 | Intron | XP_016870178.1 | ||||
XM_017014690.1 | Intron | XP_016870179.1 | ||||
XM_017014691.1 | Intron | XP_016870180.1 | ||||
XM_017014692.1 | Intron | XP_016870181.1 | ||||
XM_017014693.1 | Intron | XP_016870182.1 |