Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATTACTTTCTACGACAAAACCC[A/G]CAAATACTTTTGCACCAACCTAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600814 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR548L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR548L - microRNA 548l | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRE11A - MRE11 homolog A, double strand break repair nuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005590.3 | Intron | NP_005581.2 | ||||
NM_005591.3 | Intron | NP_005582.1 | ||||
XM_005274008.3 | Intron | XP_005274065.1 | ||||
XM_006718842.3 | Intron | XP_006718905.1 | ||||
XM_011542837.2 | Intron | XP_011541139.1 | ||||
XM_017017772.1 | Intron | XP_016873261.1 | ||||
XM_017017773.1 | Intron | XP_016873262.1 |