Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604728 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USP3 - ubiquitin specific peptidase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256702.1 | Intron | NP_001243631.1 | ||||
NM_006537.3 | Intron | NP_006528.2 | ||||
XM_017022763.1 | Intron | XP_016878252.1 | ||||
XM_017022764.1 | Intron | XP_016878253.1 | ||||
XM_017022765.1 | Intron | XP_016878254.1 | ||||
XM_017022766.1 | Intron | XP_016878255.1 | ||||
XM_017022767.1 | Intron | XP_016878256.1 | ||||
XM_017022768.1 | Intron | XP_016878257.1 |