Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTTAATTCTTAGTCCTTCACTT[C/T]GTAAACAAGGAAATCTCTTTCCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
64 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176844 MIM: 602458 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PSMA5 - proteasome subunit alpha 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199772.1 | 1586 | UTR 3 | NP_001186701.1 | |||
NM_001199773.1 | 1586 | UTR 3 | NP_001186702.1 | |||
NM_001199774.1 | 1586 | UTR 3 | NP_001186703.1 | |||
NM_002790.3 | 1586 | UTR 3 | NP_002781.2 |
SORT1 - sortilin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |