Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAAAAAGAATTATCTGTGAACCA[C/T]ACGTGATTAATAAAGATTTCCTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 112266 MIM: 616412 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLOC1S5-TXNDC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BLOC1S5-TXNDC5 - BLOC1S5-TXNDC5 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BMP6 - bone morphogenetic protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TXNDC5 - thioredoxin domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145549.2 | 2783 | UTR 3 | NP_001139021.1 | |||
NM_030810.3 | 2783 | UTR 3 | NP_110437.2 |
Set Membership: |
HapMap |