Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAATTTCTCTAGGTTGTTTGCCT[G/T]TAAGTGAAATTGATGGTTTGAAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616865 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAPOLG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PAPOLG - poly(A) polymerase gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022894.3 | Intron | NP_075045.2 | ||||
XM_005264500.3 | Intron | XP_005264557.1 | ||||
XM_005264501.1 | Intron | XP_005264558.1 | ||||
XM_011533041.1 | Intron | XP_011531343.1 | ||||
XM_017004723.1 | Intron | XP_016860212.1 | ||||
XM_017004724.1 | Intron | XP_016860213.1 |
Set Membership: |
HapMap |