Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCCTAGTCTCTGAGCTGAGTGC[C/T]GACTTAGGGAGGCAATGTTATACCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612490 MIM: 607029 MIM: 603177 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF181 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNF181 - ring finger protein 181 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VAMP5 - vesicle associated membrane protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006634.2 | Intron | NP_006625.1 |
VAMP8 - vesicle associated membrane protein 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |