Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGACATGTCACCATATTTCAGCC[C/G]TATTCAAAAATAGCCCAAAATGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616285 MIM: 612941 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMNL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FMNL2 - formin like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRPF40A - pre-mRNA processing factor 40 homolog A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017892.3 | 6750 | UTR 3 | NP_060362.3 | |||
XM_005246676.4 | 6750 | Intron | XP_005246733.3 | |||
XM_005246677.4 | 6750 | Intron | XP_005246734.2 | |||
XM_011511446.2 | 6750 | Intron | XP_011509748.1 | |||
XM_011511447.2 | 6750 | Intron | XP_011509749.1 | |||
XM_011511448.2 | 6750 | Intron | XP_011509750.1 | |||
XM_011511449.2 | 6750 | Intron | XP_011509751.1 | |||
XM_017004453.1 | 6750 | Intron | XP_016859942.1 | |||
XM_017004454.1 | 6750 | Intron | XP_016859943.1 | |||
XM_017004455.1 | 6750 | Intron | XP_016859944.1 | |||
XM_017004456.1 | 6750 | Intron | XP_016859945.1 | |||
XM_017004457.1 | 6750 | Intron | XP_016859946.1 | |||
XM_017004458.1 | 6750 | Intron | XP_016859947.1 | |||
XM_017004459.1 | 6750 | Intron | XP_016859948.1 | |||
XM_017004460.1 | 6750 | Intron | XP_016859949.1 | |||
XM_017004461.1 | 6750 | Intron | XP_016859950.1 |