Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605458 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTR8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ACTR8 - ARP8 actin-related protein 8 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022899.4 | 1358 | Intron | NP_075050.3 | |||
XM_005265587.4 | 1358 | Intron | XP_005265644.1 | |||
XM_011534249.2 | 1358 | Intron | XP_011532551.1 |
IL17RB - interleukin 17 receptor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018725.3 | 1358 | Nonsense Mutation | CAG,TAG | Q,* 484 | NP_061195.2 | |
XM_005265310.4 | 1358 | Nonsense Mutation | CAG,TAG | Q,* 513 | XP_005265367.1 | |
XM_005265311.4 | 1358 | Nonsense Mutation | CAG,TAG | Q,* 497 | XP_005265368.1 | |
XM_005265312.4 | 1358 | Nonsense Mutation | CAG,TAG | Q,* 468 | XP_005265369.1 | |
XM_011533941.2 | 1358 | UTR 3 | XP_011532243.1 | |||
XM_017006804.1 | 1358 | Nonsense Mutation | CAG,TAG | Q,* 396 | XP_016862293.1 | |
XM_017006805.1 | 1358 | Nonsense Mutation | CAG,TAG | Q,* 380 | XP_016862294.1 | |
XM_017006806.1 | 1358 | Nonsense Mutation | CAG,TAG | Q,* 367 | XP_016862295.1 | |
XM_017006807.1 | 1358 | UTR 3 | XP_016862296.1 |
Set Membership: |
HapMap Validated |