Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611949 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBOAT2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MBOAT2 - membrane bound O-acyltransferase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321265.1 | 4262 | UTR 3 | NP_001308194.1 | |||
NM_001321266.1 | 4262 | UTR 3 | NP_001308195.1 | |||
NM_001321267.1 | 4262 | UTR 3 | NP_001308196.1 | |||
NM_138799.3 | 4262 | UTR 3 | NP_620154.2 | |||
XM_011510313.1 | 4262 | Intron | XP_011508615.1 | |||
XM_011510314.2 | 4262 | Intron | XP_011508616.1 |
Set Membership: |
HapMap |