Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATGCGCCGGGATCCACAGCAGC[A/C]TGAGCTCCAAGCCCAACCAGCGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606376 | ||||||||||||||||||||
Literature Links: |
CHST10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHST10 - carbohydrate sulfotransferase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004854.4 | 2627 | UTR 3 | NP_004845.1 | |||
XM_011512207.1 | 2627 | UTR 3 | XP_011510509.1 | |||
XM_011512208.1 | 2627 | UTR 3 | XP_011510510.1 | |||
XM_011512210.1 | 2627 | UTR 3 | XP_011510512.1 | |||
XM_011512211.1 | 2627 | UTR 3 | XP_011510513.1 | |||
XM_011512212.1 | 2627 | UTR 3 | XP_011510514.1 | |||
XM_017005380.1 | 2627 | UTR 3 | XP_016860869.1 | |||
XM_017005381.1 | 2627 | UTR 3 | XP_016860870.1 | |||
XM_017005382.1 | 2627 | UTR 3 | XP_016860871.1 | |||
XM_017005383.1 | 2627 | UTR 3 | XP_016860872.1 |