Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCGATTTGGGTTTCTTCTTTTCT[A/T]CTCCTCCTTTTTAATAATACAGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602214 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSNK1G2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CSNK1G2 - casein kinase 1 gamma 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319.6 | Intron | NP_001310.3 | ||||
XM_005259498.1 | Intron | XP_005259555.1 | ||||
XM_005259499.3 | Intron | XP_005259556.1 | ||||
XM_005259500.1 | Intron | XP_005259557.1 | ||||
XM_005259501.2 | Intron | XP_005259558.1 | ||||
XM_017026297.1 | Intron | XP_016881786.1 | ||||
XM_017026298.1 | Intron | XP_016881787.1 |
CSNK1G2-AS1 - CSNK1G2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |