Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAATCATGAGGGCAGACAGGTAA[A/G]GAATCTGCCCTCATCTTTGCACTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601799 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927730 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC101927730 - uncharacterized LOC101927730 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERPINB9 - serpin family B member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004155.5 | 1517 | UTR 3 | NP_004146.1 | |||
XM_005249184.4 | 1517 | UTR 3 | XP_005249241.1 | |||
XM_011514678.2 | 1517 | UTR 3 | XP_011512980.1 | |||
XM_017010942.1 | 1517 | UTR 3 | XP_016866431.1 | |||
XM_017010943.1 | 1517 | UTR 3 | XP_016866432.1 |
Set Membership: |
HapMap Validated |