Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATATTTGTCACAGTAAGGGATTGA[C/T]ATTGGTATTACTGTGGACTAAACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603835 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFA10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NDUFA10 - NADH:ubiquinone oxidoreductase subunit A10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR6B2 - olfactory receptor family 6 subfamily B member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRR21 - proline rich 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |