Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606137 MIM: 130660 MIM: 614058 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CGREF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CGREF1 - cell growth regulator with EF-hand domain 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EMILIN1 - elastin microfibril interfacer 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KHK - ketohexokinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000221.2 | Intron | NP_000212.1 | ||||
NM_006488.2 | Intron | NP_006479.1 | ||||
XM_005264294.3 | Intron | XP_005264351.1 | ||||
XM_005264296.3 | Intron | XP_005264353.1 | ||||
XM_005264298.3 | Intron | XP_005264355.1 | ||||
XM_006712008.3 | Intron | XP_006712071.1 | ||||
XM_006712009.3 | Intron | XP_006712072.1 | ||||
XM_006712010.3 | Intron | XP_006712073.1 | ||||
XM_006712011.3 | Intron | XP_006712074.1 | ||||
XM_006712012.3 | Intron | XP_006712075.1 | ||||
XM_006712013.3 | Intron | XP_006712076.1 | ||||
XM_006712014.3 | Intron | XP_006712077.1 | ||||
XM_017004060.1 | Intron | XP_016859549.1 | ||||
XM_017004061.1 | Intron | XP_016859550.1 |
Set Membership: |
HapMap Validated |