Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607760 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOPBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TOPBP1 - topoisomerase (DNA) II binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007027.3 | 3734 | Silent Mutation | CTG,TTG | L,L 1360 | NP_008958.2 | |
XM_005247076.3 | 3734 | Silent Mutation | CTG,TTG | L,L 1355 | XP_005247133.1 | |
XM_011512357.2 | 3734 | Intron | XP_011510659.1 | |||
XM_017005636.1 | 3734 | Silent Mutation | CTG,TTG | L,L 1360 | XP_016861125.1 | |
XM_017005637.1 | 3734 | Silent Mutation | CTG,TTG | L,L 925 | XP_016861126.1 |
Set Membership: |
HapMap Validated |