Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTGCTTCCTGGGTACCTGATTC[A/G]CAGCGTACCTCACCCCGGTCCAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609185 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZHX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZHX2 - zinc fingers and homeoboxes 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014943.3 | Intron | NP_055758.1 | ||||
XM_005250836.4 | Intron | XP_005250893.1 | ||||
XM_005250837.4 | Intron | XP_005250894.1 | ||||
XM_011516931.2 | Intron | XP_011515233.1 | ||||
XM_011516932.2 | Intron | XP_011515234.1 |