Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608147 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TUBGCP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TUBGCP5 - tubulin gamma complex associated protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102610.1 | 3282 | Intron | NP_001096080.1 | |||
NM_052903.4 | 3282 | UTR 3 | NP_443135.3 | |||
XM_005272540.2 | 3282 | UTR 3 | XP_005272597.1 | |||
XM_011543868.1 | 3282 | Intron | XP_011542170.1 | |||
XM_011543869.2 | 3282 | Intron | XP_011542171.1 | |||
XM_011543870.1 | 3282 | Intron | XP_011542172.1 | |||
XM_017021890.1 | 3282 | Intron | XP_016877379.1 | |||
XM_017021891.1 | 3282 | Intron | XP_016877380.1 | |||
XM_017021892.1 | 3282 | Intron | XP_016877381.1 | |||
XM_017021893.1 | 3282 | Intron | XP_016877382.1 | |||
XM_017021894.1 | 3282 | Intron | XP_016877383.1 | |||
XM_017021895.1 | 3282 | Intron | XP_016877384.1 | |||
XM_017021896.1 | 3282 | Intron | XP_016877385.1 | |||
XM_017021897.1 | 3282 | Intron | XP_016877386.1 | |||
XM_017021898.1 | 3282 | Intron | XP_016877387.1 | |||
XM_017021899.1 | 3282 | Intron | XP_016877388.1 | |||
XM_017021900.1 | 3282 | Intron | XP_016877389.1 | |||
XM_017021901.1 | 3282 | Intron | XP_016877390.1 |
Set Membership: |
HapMap |