Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605128 MIM: 603205 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GUCA1C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GUCA1C - guanylate cyclase activator 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MORC1 - MORC family CW-type zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014429.3 | 2950 | Intron | NP_055244.3 | |||
XM_005247362.2 | 2950 | Intron | XP_005247419.1 | |||
XM_011512691.1 | 2950 | Intron | XP_011510993.1 | |||
XM_011512692.1 | 2950 | Intron | XP_011510994.1 | |||
XM_011512693.1 | 2950 | Intron | XP_011510995.1 | |||
XM_011512694.1 | 2950 | Intron | XP_011510996.1 | |||
XM_011512696.2 | 2950 | Intron | XP_011510998.1 | |||
XM_011512697.2 | 2950 | Intron | XP_011510999.1 | |||
XM_017006169.1 | 2950 | Missense Mutation | GCC,GTC | A,V 969 | XP_016861658.1 | |
XM_017006170.1 | 2950 | Intron | XP_016861659.1 |