Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600239 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPR1 - G protein-coupled receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098199.1 | 755 | Silent Mutation | GAC,GAT | D,D 177 | NP_001091669.1 | |
NM_001261452.1 | 755 | Silent Mutation | GAC,GAT | D,D 177 | NP_001248381.1 | |
NM_001261453.1 | 755 | Silent Mutation | GAC,GAT | D,D 177 | NP_001248382.1 | |
NM_001261454.1 | 755 | Silent Mutation | GAC,GAT | D,D 177 | NP_001248383.1 | |
NM_001261455.1 | 755 | Silent Mutation | GAC,GAT | D,D 177 | NP_001248384.1 | |
NM_005279.3 | 755 | Silent Mutation | GAC,GAT | D,D 177 | NP_005270.2 | |
XM_005246471.2 | 755 | Silent Mutation | GAC,GAT | D,D 177 | XP_005246528.1 | |
XM_017003832.1 | 755 | Silent Mutation | GAC,GAT | D,D 177 | XP_016859321.1 |
Set Membership: |
HapMap |