Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603758 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GSTZ1 - glutathione S-transferase zeta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMED8 - transmembrane p24 trafficking protein family member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_213601.1 | 7337 | Intron | NP_998766.1 | |||
XM_017021223.1 | 7337 | UTR 3 | XP_016876712.1 | |||
XM_017021224.1 | 7337 | UTR 3 | XP_016876713.1 | |||
XM_017021225.1 | 7337 | UTR 3 | XP_016876714.1 |