Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACATGTTCTCTCATTTAAGAGCA[C/T]GTGTCTTTTATGCGTGTTAGGCATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300390 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHM PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CHM - CHM, Rab escort protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000390.3 | Intron | NP_000381.1 | ||||
NM_001145414.3 | Intron | NP_001138886.1 | ||||
NM_001320959.1 | Intron | NP_001307888.1 | ||||
XM_017029242.1 | Intron | XP_016884731.1 | ||||
XM_017029243.1 | Intron | XP_016884732.1 | ||||
XM_017029244.1 | Intron | XP_016884733.1 | ||||
XM_017029245.1 | Intron | XP_016884734.1 | ||||
XM_017029246.1 | Intron | XP_016884735.1 |
Set Membership: |
HapMap |