Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCATACTGAACCTGACTCTCAG[C/T]GAGACTTTGCTGGCCCTGAGAATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300197 MIM: 300104 MIM: 300394 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6AP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ATP6AP1 - ATPase H+ transporting accessory protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001183.5 | Intron | NP_001174.2 | ||||
XM_011531179.1 | Intron | XP_011529481.1 |
CH17-340M24.3 - uncharacterized protein BC009467 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GDI1 - GDP dissociation inhibitor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAZ - tafazzin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |