Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603443 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HCG25 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HCG25 - HLA complex group 25 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS52 - VPS52, GARP complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289174.1 | 2779 | UTR 3 | NP_001276103.1 | |||
NM_001289175.1 | 2779 | UTR 3 | NP_001276104.1 | |||
NM_001289176.1 | 2779 | UTR 3 | NP_001276105.1 | |||
NM_022553.5 | 2779 | UTR 3 | NP_072047.4 | |||
XM_011514797.1 | 2779 | UTR 3 | XP_011513099.1 | |||
XM_011514798.2 | 2779 | UTR 3 | XP_011513100.1 | |||
XM_011514799.1 | 2779 | UTR 3 | XP_011513101.1 | |||
XM_017011177.1 | 2779 | Intron | XP_016866666.1 | |||
XM_017011178.1 | 2779 | UTR 3 | XP_016866667.1 | |||
XM_017011179.1 | 2779 | Intron | XP_016866668.1 |
Set Membership: |
HapMap |