Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608187 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCM8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MCM8 - minichromosome maintenance 8 homologous recombination repair factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281520.1 | 176 | UTR 5 | NP_001268449.1 | |||
NM_001281521.1 | 176 | UTR 5 | NP_001268450.1 | |||
NM_001281522.1 | 176 | UTR 5 | NP_001268451.1 | |||
NM_032485.5 | 176 | UTR 5 | NP_115874.3 | |||
NM_182802.2 | 176 | UTR 5 | NP_877954.1 | |||
XM_011529387.2 | 176 | UTR 5 | XP_011527689.1 | |||
XM_017028105.1 | 176 | UTR 5 | XP_016883594.1 | |||
XM_017028106.1 | 176 | UTR 5 | XP_016883595.1 | |||
XM_017028107.1 | 176 | Intron | XP_016883596.1 |
TRMT6 - tRNA methyltransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281467.1 | 176 | Intron | NP_001268396.1 | |||
NM_015939.4 | 176 | Intron | NP_057023.2 | |||
XM_017027875.1 | 176 | Intron | XP_016883364.1 |
Set Membership: |
HapMap |