Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTTAATTTTGGTGATGTGACTAT[A/G]GAAGCTGTACTTTACACCTTTATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603455 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929709 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC101929709 - uncharacterized LOC101929709 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RIPK2 - receptor interacting serine/threonine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003821.5 | Intron | NP_003812.1 | ||||
XM_005251092.3 | Intron | XP_005251149.1 | ||||
XM_011517357.2 | Intron | XP_011515659.1 |
Set Membership: |
HapMap Validated |