Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCAAGAAGGAATGAGGAGAGGAT[A/G]TGAGGAAAGAGCCACAGCACAGAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRHR1-IT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CRHR1-IT1 - CRHR1 intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MGC57346 - ADP-ribosylation factor pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MGC57346-CRHR1 - MGC57346-CRHR1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256299.2 | Intron | NP_001243228.1 | ||||
NM_001303016.1 | Intron | NP_001289945.1 |
Set Membership: |
HapMap |