Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTAAGCTTAGGGGGCTTGTTGA[A/G]CATGTTCTGTGGTTCTGTGTGCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608329 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DKFZP434K028 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DKFZP434K028 - uncharacterized LOC26070 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100129473 - uncharacterized LOC100129473 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYRF - myelin regulatory factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127392.2 | Intron | NP_001120864.1 | ||||
NM_013279.3 | Intron | NP_037411.1 | ||||
XM_005274222.1 | Intron | XP_005274279.1 | ||||
XM_005274223.1 | Intron | XP_005274280.1 | ||||
XM_005274224.1 | Intron | XP_005274281.1 | ||||
XM_005274225.1 | Intron | XP_005274282.1 | ||||
XM_005274226.1 | Intron | XP_005274283.1 | ||||
XM_005274227.1 | Intron | XP_005274284.1 | ||||
XM_005274228.1 | Intron | XP_005274285.1 | ||||
XM_011545234.2 | Intron | XP_011543536.1 |