Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601825 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MUM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MUM1 - melanoma associated antigen (mutated) 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFS7 - NADH:ubiquinone oxidoreductase core subunit S7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024407.4 | 80 | Intron | NP_077718.3 | |||
XM_005259556.4 | 80 | Intron | XP_005259613.2 | |||
XM_017026768.1 | 80 | UTR 5 | XP_016882257.1 |
Set Membership: |
HapMap |